1-(4-Aminophenyl)-4-methylpiperazine

Leverantör: Thermo Scientific

Synonyms: 4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline

SS00001DA SS00001EA
MAYBSS00001DAEA 0 SEK
MAYBSS00001DA MAYBSS00001EA
1-(4-Aminophenyl)-4-methylpiperazine
1-(4-Aminophenyl)-4-methylpiperazine
Formel: C₁₁H₁₇N₃
MW: 191,28 g/mol
MDL Number: MFCD00172703
CAS nummer: 16153-81-4
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR