Ethyl 3,5-dichloro-4-hydroxybenzoate

Leverantör: Thermo Scientific

Synonyms: Ethyl 3,5-dichloro-4-hydroxybenzoate

SPB00157EA
MAYBSPB00157EAEA 0 SEK
MAYBSPB00157EA
Ethyl 3,5-dichloro-4-hydroxybenzoate
Ethyl-3,5-dichloro-4-hydroxybenzoate
Formel: C₉H₈Cl₂O₃
MW: 235,07 g/mol
MDL Number: MFCD00016420
CAS nummer: 17302-82-8
EINECS: 241-331-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR