2-Methyl-5-phenyl-3-furoic acid

Leverantör: Thermo Scientific

Synonyms: 2-Methyl-5-phenylfuran-3-carboxylic acid

CC14901DA CC14901EA
MAYBCC14901DAEA 0 SEK
MAYBCC14901DA MAYBCC14901EA
2-Methyl-5-phenyl-3-furoic acid
2-Methyl-5-phenyl-3-furoic acid
Formel: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD00221070
CAS nummer: 108124-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR