Methyl-3-methylcrotonate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3,3-dimethylacrylate, Methyl-3,3-dimethylacrylate
L19731.09 L19731.18
L19731.09EA 1600 SEK
L19731.09 L19731.18
Methyl-3-methylcrotonate ≥98%
Methyl-3-methylcrotonate
Purity: 98% CAS No.: 924-50-5 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 33°(91°F) Refractive Index: 1.4375 Melting Point: -41° Boiling Point: 134-139° Beilstein Reference: 1741592 UN No.: 3272 MDL No.: MFCD00043940
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 134…139 °C
Smältpunkt: –41 °C
Densitet: 0,936
Flampunkt: 33 °C (91 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00043940
CAS nummer: 924-50-5
EINECS: 213-107-4
UN: 3272
ADR: 3,III