Bismuth(III) trifluoromethanesulphonate ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Bismuth triflate, Bismuth(III) trifluoromethanesulphonate
L19687.06 L19687.14
L19687.06EA 1575 SEK
L19687.06 L19687.14
Bismuth(III) trifluoromethanesulphonate ≥99%
Bismuth(III) trifluoromethanesulphonate
Purity: 99% CAS No.: 88189-03-1 Molecular Formula: C3BiF9O9S3 Formula Weight: 656.19 Melting Point: 300° UN No.: 3261 MDL No.: MFCD02093669
Formel: C₃BiF₉O₉S₃
MW: 656,19 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD02093669
CAS nummer: 88189-03-1
UN: 3261
ADR: 8,III