1-Allylpiperazine ≥98%

Leverantör: Thermo Scientific
Fara

L19391.06 L19391.14
L19391.06EA 2025 SEK
L19391.06 L19391.14
1-Allylpiperazine ≥98%
1-Allylpiperazine

Purity: 98+% CAS No.: 13961-36-9 Molecular Formula: C7H14N2 Formula Weight: 126.20 Flash Point: 51°(123°F) Refractive Index: 1.4830 Boiling Point: 180-182° Beilstein Reference: 507447 UN No.: 2920 MDL No.: MFCD00167970

Formel: C₇H₁₄N₂
MW: 126,2 g/mol
Kokpunkt: 180…182 °C
Densitet: 0,902
Flampunkt: 51 °C (123 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00167970
CAS nummer: 13961-36-9
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR