Methyl-3,5-difluorobenzoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3,5-difluorobenzoate
L17171.06 L17171.14
L17171.06EA 1300 SEK
L17171.06 L17171.14
Methyl-3,5-difluorobenzoate ≥98%
Methyl-3,5-difluorobenzoate
Purity: 98% CAS No.: 216393-55-4 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Refractive Index: 1.4740
Formel: C₈H₆F₂O₂
MW: 172,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01318160
CAS nummer: 216393-55-4