Methyl-2,6-difluorobenzoate ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 2,6-difluorobenzoate
L08063.06 L08063.14
L08063.06EA 985 SEK
L08063.06 L08063.14
Methyl-2,6-difluorobenzoate ≥97%
Methyl-2,6-difluorobenzoate
Purity: 97% CAS No.: 13671-00-6 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Flash Point: 95°(203°F) Refractive Index: 1.4760 Boiling Point: 203-204° Beilstein Reference: 1948618 MDL No.: MFCD00051778
Formel: C₈H₆F₂O₂
MW: 172,13 g/mol
Kokpunkt: 203…204 °C
Densitet: 1,281
Flampunkt: 95 °C (203 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051778
CAS nummer: 13671-00-6