2-Formyl-6-methoxypyridine 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde
H34432.03 H34432.06
H34432.03EA 1500 SEK
H34432.03 H34432.06
2-Formyl-6-methoxypyridine 96%
2-Formyl-6-methoxypyridine
Purity: 96% CAS No.: 54221-96-4 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Refractive Index: 1.5310
Formel: C₇H₇NO₂
MW: 137.14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05664030
CAS nummer: 54221-96-4