1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: PYR14-TFSI, N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imide
H27177.06 H27177.18
H27177.06EA 1750 SEK
H27177.06 H27177.18
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide ≥98%
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide
Purity: 98% CAS No.: 223437-11-4 Molecular Formula: C11H20F6N2O4S2 Formula Weight: 422.42 MDL No.: MFCD07784447
Formel: C₁₁H₂₀F₆N₂O₄S₂
MW: 422.41 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07784447
CAS nummer: 223437-11-4