trans-2,5-Dimethylpiperazine ≥98%
Leverantör: Thermo Scientific
Fara
B23150.06 B23150.14 B23150.22
B23150.06EA 900 SEK
B23150.06 B23150.14 B23150.22
trans-2,5-Dimethylpiperazine ≥98%
trans-2,5-Dimethylpiperazine
Purity: 98% CAS No.: 2815-34-1 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 58°(136°F) Melting Point: 115-119° Boiling Point: 162-165° Beilstein Reference: 79886 UN No.: 2925 MDL No.: MFCD00070516
Formel: C₆H₁₄N₂
MW: 114,19 g/mol
Kokpunkt: 162…165 °C
Smältpunkt: 115…119 °C
Flampunkt: 58 °C (136 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00070516
CAS nummer: 2815-34-1
ADR: 4.1,III