Methyl-2,5-dichlorobenzoate ≥99%
Leverantör: Thermo Scientific
Synonyms: Methyl 2,5-dichlorobenzoate
B22489.09 B22489.18
B22489.09EA 1300 SEK
B22489.09 B22489.18
Methyl-2,5-dichlorobenzoate ≥99%
Methyl-2,5-dichlorobenzoate
Purity: 99% CAS No.: 2905-69-3 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Flash Point: 110°(230°F) Melting Point: 37-40° Beilstein Reference: 2251582 MDL No.: MFCD00000606
Formel: C₈H₆Cl₂O₂
MW: 205,04 g/mol
Smältpunkt: 37…40 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000606
CAS nummer: 2905-69-3
EINECS: 220-815-7