DL-Tropic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid
B22040.09 B22040.18
B22040.09EA 750 SEK
B22040.09 B22040.18
DL-Tropic acid ≥98%
DL-Tropic acid
Purity: 98% CAS No.: 552-63-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 117-119° Beilstein Reference: 2209199 Merck Reference: 14,9779 MDL No.: MFCD00004255
Formel: C₉H₁₀O₃
MW: 166.18 g/mol
Smältpunkt: 117…119 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004255
CAS nummer: 529-64-6
EINECS: 209-020-6