TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
B21819.09 B21819.18 B21819.30
B21819.09EA 810 SEK
B21819.09 B21819.18 B21819.30
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid) ≥99%
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Purity: 99% CAS No.: 7365-44-8 Molecular Formula: C6H15NO6S Formula Weight: 229.25 Odour: Odourless Melting Point: ca 225° dec. Beilstein Reference: 1957061 Merck Reference: 14,9176 MDL No.: MFCD00007532
Formel: C₆H₁₅NO₆S
MW: 229,25 g/mol
Smältpunkt: ∼ 225 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007532
CAS nummer: 7365-44-8
EINECS: 230-906-3
Merck Index: 14,9176