1-Acetylpiperazine 98%
Leverantör: Thermo Scientific
Varning
B20200.06 B20200.14 B20200.22
B20200.06EA 845 SEK
B20200.06 B20200.14 B20200.22
1-Acetylpiperazine 98%
1-Acetylpiperazine
Purity: 99% CAS No.: 13889-98-0 Molecular Formula: C6H12N2O Formula Weight: 128.18 Flash Point: 110°(230°F) Melting Point: 30-34° Boiling Point: 127-130°/8mm Beilstein Reference: 112220 MDL No.: MFCD00058676
Formel: C₆H₁₂N₂O
MW: 128,17 g/mol
Kokpunkt: 127…130 °C (8 mmHg)
Smältpunkt: 30…34 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00058676
CAS nummer: 13889-98-0
EINECS: 237-659-0