Ethyl vinyl ether ≥99% stabilised
Leverantör: Thermo Fisher Scientific
Fara
Stabilization: Stabilised with N,N-diethylaniline
A15691.0F A15691.AK A15691.AP
A15691.0FEA 3775 SEK
A15691.0F A15691.AK A15691.AP
Ethyl vinyl ether ≥99% stabilised
Ethyl vinyl ether
Stabilized
Purity: 99% CAS No.: 109-92-2 Molecular Formula: C4H8O Formula Weight: 72.11 Storage Temperature: 2°C to 8°C Flash Point: -45°(-49°F) Refractive Index: 1.3765 Odour: Ether-like Melting Point: -115° Boiling Point: 33-34° Beilstein Reference: 605351 UN No.: 1302 MDL No.: MFCD00009248
Formel: C₄H₈O
MW: 72.11 g/mol
Kokpunkt: 33…34 °C
Smältpunkt: –115 °C
Densitet: 0.754
Flampunkt: –45 °C (–49 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00009248
CAS nummer: 109-92-2
EINECS: 203-718-4
UN: 1302
ADR: 3 I