Ethyl-2-furoate ≥99%
Leverantör: Thermo Scientific
Synonyms: Ethyl2-furoate, Ethyl furan-2-carboxylate
A13909.14 A13909.22 A13909.36
A13909.14EA 570 SEK
A13909.14 A13909.22 A13909.36
Ethyl-2-furoate ≥99%
Ethyl-2-furoate
Purity: 99% CAS No.: 614-99-3 Molecular Formula: C7H8O3 Formula Weight: 140.14 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Melting Point: 31-36° Boiling Point: 195-196° Beilstein Reference: 2653 Merck Reference: 14,4307 MDL No.: MFCD00003237
Formel: C₇H₈O₃
MW: 140,14 g/mol
Kokpunkt: 195…196 °C
Smältpunkt: 31…36 °C
Densitet: 1,120
Flampunkt: 70 °C (158 °F)
MDL Number: MFCD00003237
CAS nummer: 614-99-3
EINECS: 210-404-0
Merck Index: 14,4307