3-Picolyl chloride hydrochloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride
A13279.06 A13279.14
A13279.06EA 735 SEK
A13279.06 A13279.14
3-Picolyl chloride hydrochloride ≥97%
3-Picolyl chloride hydrochloride
Purity: 97% CAS No.: 6959-48-4 Molecular Formula: C6H6ClN.HCl Formula Weight: 164.04 pH: 3 Melting Point: 136-142° Beilstein Reference: 3689156 UN No.: 3261 MDL No.: MFCD00012818
Formel: C₆H₇Cl₂N
MW: 164,03 g/mol
Smältpunkt: 136…142 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012818
CAS nummer: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III