Methyl-2-methyl-3-furoate ≥97%
Leverantör: Thermo Fisher Scientific
Varning
A11979.14 A11979.22
A11979.14EA 1700 SEK
A11979.14 A11979.22
Methyl-2-methyl-3-furoate ≥97%
Methyl-2-methyl-3-furoate
Purity: 97% CAS No.: 6141-58-8 Molecular Formula: C7H8O3 Formula Weight: 140.14 Flash Point: 63°(145°F) Refractive Index: 1.4745 Boiling Point: 71-72°/20mm Beilstein Reference: 1342040 MDL No.: MFCD00003249
Formel: C₇H₈O₃
Kokpunkt: 71…72 °C (20 mmHg)
Densitet: 1,114
Flampunkt: 63 °C (145 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003249
CAS nummer: 6141-58-8
EINECS: 228-132-6