2-Picolyl chloride hydrochloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-(Chloromethyl)pyridine HCl, 2-(Chloromethyl)pyridine hydrochloride
A10226.14 A10226.06 A10226.22
A10226.14EA 2750 SEK
A10226.14 A10226.06 A10226.22
2-Picolyl chloride hydrochloride ≥98%
2-Picolyl chloride hydrochloride
Purity: 98% CAS No.: 6959-47-3 Molecular Formula: C6H6ClN.HCl Formula Weight: 164.04 Melting Point: 120-124° Beilstein Reference: 3689155 UN No.: 3261 MDL No.: MFCD00012811
Formel: C₆H₇Cl₂N
MW: 164.03 g/mol
Smältpunkt: 120…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012811
CAS nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8 III