Litiumbis(trifluorometylsulfonyl)imid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt
381030100 381030500 381030000
ACRO381030100EA 820 SEK
ACRO381030100 ACRO381030500 381030000.
Litiumbis(trifluorometylsulfonyl)imid 99%
Litiumbis(trifluorometylsulfonyl)imid
Formel: C₂F₆LiNO₄S₂
MW: 287,09 g/mol
Smältpunkt: 234…238 °C
Storage Temperature: Omgivande
MDL Number: MFCD00210017
CAS nummer: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms
Titration after Ion exchange ≥98.5 %