1-Acetylpiperazine 98%
Leverantör: Thermo Scientific
Varning
346830050 346830250
ACRO346830050EA 725 SEK
ACRO346830050 ACRO346830250
1-Acetylpiperazine 98%
1-Acetylpiperazine
Formel: C₆H₁₂N₂O
MW: 128,17 g/mol
Smältpunkt: 31…34 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00058676
CAS nummer: 13889-98-0
EINECS: 237-659-0
Order Now
Specifikation testresultat
Appearance Clear light yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %