Chloro(diisopropyl)phosphine 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Di-i-propylchlorophosphine
321550050 321550250
ACRO321550050EA 1275 SEK
ACRO321550050 ACRO321550250
Chloro(diisopropyl)phosphine 96%
Chloro(diisopropyl)phosphine
Formel: C₆H₁₄ClP
MW: 152.6 g/mol
Kokpunkt: 69 °C (33 mmHg)
Densitet: 0.95 g/cm³
Flampunkt: 3 °C
Storage Temperature: Omgivande
MDL Number: MFCD00015027
CAS nummer: 40244-90-4
UN: 2924
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
Titration Argentometric ≥95 %