(±)-α-Terpineol 97+%
Leverantör: Thermo Scientific
Varning
Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
301610250 301615000 301610025
ACRO301610250EA 505 SEK
ACRO301610250 ACRO301615000 ACRO301610025
(±)-α-Terpineol 97+%
(±)-α-Terpineol
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokpunkt: 214…224 °C (760 mmHg)
Smältpunkt: 31…34 °C
Densitet: 0,937 g/cm³
Flampunkt: 90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001557
CAS nummer: 98-55-5
EINECS: 202-680-6
Order Now
Specifikation testresultat
Appearance Clear colorless to white solid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4850 (20°C, 589 nm)
GC ≥97.0 %
Specific gravity (20°C) 0.934 to 0.940