2-Aminoterephthalic acid 99%
Leverantör: Thermo Scientific
Varning
278030250 278031000
ACRO278030250EA 1200 SEK
ACRO278030250 ACRO278031000
2-Aminoterephthalic acid 99%
2-Aminoterephthalic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: 324 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00134536
CAS nummer: 10312-55-7
Order Now
Specifikation testresultat
Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %