Methyl-3-bromopropionate 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Methyl 3-bromopropanoate
221790500 221791000 221790250
ACRO221790500EA 2500 SEK
ACRO221790500 ACRO221791000 ACRO221790250
Methyl-3-bromopropionate 97%
Methyl-3-bromopropionate
Formel: C₄H₇BrO₂
Kokpunkt: 64…66 °C (18 mmHg)
Densitet: 1,53 g/cm³
Flampunkt: 75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000250
CAS nummer: 3395-91-3
EINECS: 222-247-5
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4565 to 1.4595 (20°C, 589 nm)
GC ≥96.0 %