3-Furoic acid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid
172450250 172450050
ACRO172450250EA 4650 SEK
ACRO172450250 ACRO172450050
3-Furoic acid 99%
3-Furoic acid
Formel: C₅H₄O₃
MW: 112,08 g/mol
Smältpunkt: 121…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005350
CAS nummer: 488-93-7
EINECS: 207-689-9
Order Now
Specifikation testresultat
Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 124 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %