Tropinone 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one
140570100 140570050 140570500
ACRO140570100EA 1250 SEK
ACRO140570100 ACRO140570050 ACRO140570500
Tropinone 99%
Tropinone
Formel: C₈H₁₃NO
MW: 139.2 g/mol
Kokpunkt: 113 °C (25 mmHg)
Smältpunkt: 41…44 °C
Flampunkt: 90 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005549
CAS nummer: 532-24-1
EINECS: 208-530-6
UN: 3259
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Light yellow to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 44 °C
GC ≥98.5 %