Nicotinaldehyde 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde
131830250 131831000 131835000
ACRO131830250EA 560 SEK
ACRO131830250 ACRO131831000 ACRO131835000
Nicotinaldehyde 98%
Nicotinaldehyde
Formel: C₆H₅NO
MW: 107,11 g/mol
Kokpunkt: 95…97 °C (15 mmHg)
Densitet: 1,142 g/cm³
Flampunkt: 84 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006382
CAS nummer: 500-22-1
EINECS: 207-900-4
UN: 3265
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %
pH 7.0 to 9.0
Titration with HClO4 94 to 106 %