Homophthalic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(Carboxymethyl)benzoic acid
120840250 120841000
ACRO120840250EA 625 SEK
ACRO120840250 ACRO120841000
Homophthalic acid 98%
Homophthalic acid
Formel: C₉H₈O₄
MW: 180,16 g/mol
Smältpunkt: 178…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004326
CAS nummer: 89-51-0
EINECS: 201-913-9
Order Now
Specifikation testresultat
Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %