Piperidine-N-carbaldehyde 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Formylpiperidine, N-Formylpiperidine
119730010 119732500
ACRO119730010EA 4875 SEK
ACRO119730010 ACRO119732500
Piperidine-N-carbaldehyde 99%
Piperidine-N-carbaldehyde
Formel: C₆H₁₁NO
MW: 113,16 g/mol
Smältpunkt: –31 °C
Densitet: 1,019 g/cm³
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006483
CAS nummer: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure