Cyclopropylamine 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Aminocyclopropane
263681000 263680025 263680250 263685000
ACRO263681000EA 4200 SEK
ACRO263681000 ACRO263680025 ACRO263680250 ACRO263685000
Cyclopropylamine 98%
Cyclopropylamine
Formel: C₃H₇N
MW: 57.1 g/mol
Kokpunkt: 49…50 °C (760 mmHg)
Smältpunkt: –50 °C
Densitet: 0.824 g/cm³
Flampunkt: –25 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001301
CAS nummer: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %