Cyclopentylamine 99+%
Leverantör: Thermo Fisher Scientific
Fara
111590050 111590250 111591000 111595000
ACRO111590050EA 325 SEK
ACRO111590050 ACRO111590250 ACRO111591000 ACRO111595000
Cyclopentylamine 99+%
Cyclopentylamine
Formel: C₅H₁₁N
Kokpunkt: 106…108 °C (760 mmHg)
Densitet: 0,86 g/cm³
Flampunkt: 17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001380
CAS nummer: 1003-03-8
EINECS: 213-697-3
UN: 3286
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4482 to 1.4502 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.0 %
Color scale ≤50 APHA