Chlorocyclohexane 98%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Cyclohexyl chloride
111300010
111301000
111302500
ACRO111300010EA
1975
SEK
ACRO111300010
ACRO111301000
ACRO111302500
Chlorocyclohexane 98%
Chlorocyclohexane
Formel:
C₆H₁₁Cl MW: 118,61 g/mol Kokpunkt: 142 °C (1013 hPa) Smältpunkt: –44 °C Densitet: 1 g/cm³ Flampunkt: 28 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00003822 CAS nummer: 542-18-7 EINECS: 208-806-6 UN: 1993 ADR: 3,III Merck Index: 13,02761 |
Specifikation testresultat
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥97.5 % |
Color scale | ≤10 APHA |
Water soluble titr. acid | ≤0.0005 meq/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...