Cyclopentanecarboxylic acid ≥99%
Leverantör: Thermo Fisher Scientific
Varning
A12375.09 A12375.18 A12375.30
A12375.09EA 1075 SEK
A12375.09 A12375.18 A12375.30
Cyclopentanecarboxylic acid ≥99%
Cyclopentanecarboxylic acid
Purity: 99% CAS No.: 3400-45-1 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 93°(199°F) Refractive Index: 1.4540 Melting Point: -7° Boiling Point: 215-216° Beilstein Reference: 2039729 MDL No.: MFCD00001371
Formel: C₆H₁₀O₂
Kokpunkt: 215…216 °C
Smältpunkt: –7 °C
Densitet: 1,052
Flampunkt: 93 °C (199 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001371
CAS nummer: 3400-45-1
EINECS: 222-269-5