Cyclobutanecarboxylic acid 98%
Leverantör: Thermo Scientific
Varning
110900050 110900250 110901000
ACRO110900050EA 530 SEK
ACRO110900050 ACRO110900250 ACRO110901000
Cyclobutanecarboxylic acid 98%
Cyclobutanecarboxylic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 195 °C (760 mmHg)
Smältpunkt: –20 °C
Densitet: 1,04 g/cm³
Flampunkt: 83 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001323
CAS nummer: 3721-95-7
EINECS: 223-072-7
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %