Cyclobutanone ≥98% stabilised
Leverantör: Thermo Scientific
Fara
Stabilization: Stabilised med BHT/Ionol
A13068.06 A13068.14 A13068.03
A13068.06EA 4350 SEK
A13068.06 A13068.14 A13068.03
Cyclobutanone ≥98% stabilised
Cyclobutanone
Stabilized with ca 0.01% BHT
Purity: 98% CAS No.: 1191-95-3 Molecular Formula: C4H6O Formula Weight: 70.09 Flash Point: 10°(50°F) Refractive Index: 1.4210 Odour: Pleasant Melting Point: -51° Boiling Point: 97-99° Beilstein Reference: 1560289 UN No.: 1224 MDL No.: MFCD00001332
Formel: C₄H₆O
MW: 70,09 g/mol
Kokpunkt: 97…99 °C
Smältpunkt: –51 °C
Densitet: 0,935
Flampunkt: 10 °C (50 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001332
CAS nummer: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II