Bromocyclobutane 97%, ren
Leverantör: Thermo Scientific
Fara
Synonyms: Cyclobutyl bromide
320500010 320500050 320500250
ACRO320500010EA 1250 SEK
ACRO320500010 ACRO320500050 ACRO320500250
Bromocyclobutane 97%, ren
Bromocyclobutane
Formel: C₄H₇Br
MW: 135 g/mol
Kokpunkt: 108 °C
Densitet: 1,43 g/cm³
Flampunkt: 22 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001317
CAS nummer: 4399-47-7
EINECS: 224-530-9
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4810 (20°C, 589 nm)
GC ≥96.0 %