8-Methylquinoline 97%
Leverantör: Thermo Scientific
Varning
303620250 303620050
ACRO303620250EA 3100 SEK
ACRO303620250 ACRO303620050
8-Methylquinoline 97%
8-Methylquinoline
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokpunkt: 248 °C (1013 hPa)
Smältpunkt: –80 °C
Densitet: 1,05 g/cm³
Flampunkt: 104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006810
CAS nummer: 611-32-5
EINECS: 210-264-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6170 (20°C, 589 nm)
GC ≥96.0 %