Quinaldin (2-Metylkinolin) 95+%
Leverantör: Thermo Fisher Scientific
Varning
132172500 132170010
ACRO132172500EA 1475 SEK
ACRO132172500 ACRO132170010
Quinaldin (2-Metylkinolin) 95+%
Quinaldin (2-Metylkinolin)
Formel: C₁₀H₉N
Kokpunkt: 248 °C (760 mmHg)
Smältpunkt: –2 °C
Densitet: 1,058 g/cm³
Flampunkt: 79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006756
CAS nummer: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228
Order Now
Specifikation testresultat
Appearance Clear light yellow to brown-yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6130 (20°C, 589 nm)
GC ≥95.0 %
Hydroxyl value 25 to 32 mg KOH/g