6-Quinolinol 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Quinolin-6-ol, 6-Hydroxyquinoline
364930250 364931000
ACRO364930250EA 2075 SEK
ACRO364930250 ACRO364931000
6-Quinolinol 98%
6-Quinolinol
Formel: C₉H₇NO
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: 190…195 °C
Storage Temperature: Omgivande
MDL Number: MFCD00047611
CAS nummer: 580-16-5
EINECS: 209-454-6
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Beige to grey to brown Powder
Infrared spectrum Conforms
Melting point 190 to 195 °C
HPLC ≥97.5 %