Methyl-6-quinolyl ether 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Methoxyquinoline
179340250 179341000
ACRO179340250EA 2125 SEK
ACRO179340250 ACRO179341000
Methyl-6-quinolyl ether 98%
Methyl-6-quinolyl ether
Formel: C₁₀H₉NO
MW: 159,19 g/mol
Kokpunkt: 193 °C (50 mmHg)
Smältpunkt: 18…20 °C
Densitet: 1,152 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006800
CAS nummer: 5263-87-6
EINECS: 226-077-2
Order Now
Specifikation testresultat
Appearance Clear yellow to pink to purple to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.6220 to 1.6260 (20°C, 589 nm)
GC ≥97.5 %