3-Aminoquinoline ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Quinolin-3-amine, Quinolin-3-ylamine
A0418-25G
A0418-5G
TCIAA0418-25GEA
1900
SEK
TCIAA0418-25G
TCIAA0418-5G
3-Aminoquinoline ≥99.0% (by GC, titration analysis)
3-Aminoquinoline
Formel:
C₉H₈N₂ MW: 144,18 g/mol Kokpunkt: 137°C Smältpunkt: 84 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006772 CAS nummer: 580-17-6 EINECS: 209-455-1 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 92.0 to 96.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...