3-Bromoquinoline ≥98%
Leverantör: Thermo Scientific
Varning
A14573.09 A14573.18 A14573.22
A14573.09EA 1050 SEK
A14573.09 A14573.18 A14573.22
3-Bromoquinoline ≥98%
3-Bromoquinoline
Purity: 98% CAS No.: 5332-24-1 Molecular Formula: C9H6BrN Formula Weight: 208.06 Flash Point: 110°(230°F) Refractive Index: 1.6640 Melting Point: 13-15° Boiling Point: 274-276° Beilstein Reference: 112939 MDL No.: MFCD00006767
Formel: C₉H₆BrN
MW: 208,06 g/mol
Kokpunkt: 274…276 °C
Smältpunkt: 13…15 °C
Densitet: 1,533
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006767
CAS nummer: 5332-24-1
EINECS: 226-237-1