4-Phenylbenzoic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
A12438.14 A12438.22 A12438.36
A12438.14EA 965 SEK
A12438.14 A12438.22 A12438.36
4-Phenylbenzoic acid ≥98%
4-Phenylbenzoic acid
Purity: 98% CAS No.: 92-92-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Odour: Odourless Melting Point: 223-227° Beilstein Reference: 973519 MDL No.: MFCD00002553
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Kokpunkt: 373 °C (1013 hPa)
Smältpunkt: 223…227 °C
Densitet: 1,284 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002553
CAS nummer: 92-92-2
EINECS: 202-203-1