Benzylbenzoat ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Benzoic acid benzyl ester
L03258.30 L03258.0B L03258.0E
L03258.30EA 600 SEK
L03258.30 L03258.0B L03258.0E
Benzylbenzoat ≥99%
Benzylbenzoat
Purity: 99+% CAS No.: 120-51-4 Molecular Formula: C14H12O2 Formula Weight: 212.25 Flash Point: 147°(296°F) Refractive Index: 1.5680 Melting Point: 18-20° Boiling Point: 323-324° Beilstein Reference: 2049280 Merck Reference: 14,1127 UN No.: 3082 MDL No.: MFCD00003075
Formel: C₁₄H₁₂O₂
MW: 212,25 g/mol
Kokpunkt: 323…324 °C
Smältpunkt: 18…20 °C
Densitet: 1,112
Flampunkt: 147 °C (296 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003075
CAS nummer: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 14,1127