1-Bromo-4-cyclohexylbenzene ≥98%
Leverantör: Thermo Fisher Scientific
L04636.14
L04636.06
L04636.14EA
6925
SEK
L04636.14
L04636.06
1-Bromo-4-cyclohexylbenzene ≥98%
1-Bromo-4-cyclohexylbenzene
Purity: 98%
CAS No.: 25109-28-8
Molecular Formula: C12H15Br
Formula Weight: 239.16
Refractive Index: 1.5595
Boiling Point: 90-94°/0.01mm
MDL No.: MFCD00019339
Formel:
C₁₂H₁₅Br MW: 239.16 g/mol Kokpunkt: 90…94 °C (0.01 mmHg) Densitet: 1.287 Storage Temperature: Omgivande |
MDL Number:
MFCD00019339 CAS nummer: 25109-28-8 EINECS: 246-623-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...