2-Chloromethyl-p-xylene ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,5-Dimethylbenzyl chloride, α-Chloro-1,2,5-trimethylbenzene, 2-(Chloromethyl)-1,4-dimethylbenzene, 1-(Chloromethyl)-2,5-dimethylbenzene
L01303.06 L01303.14
L01303.06EA 995 SEK
L01303.06 L01303.14
2-Chloromethyl-p-xylene ≥98%
2-Chloromethyl-p-xylene
Purity: 98% CAS No.: 824-45-3 Molecular Formula: C9H11Cl Formula Weight: 154.64 Flash Point: 65°(149°F) Refractive Index: 1.5370 Odour: Pungent Boiling Point: 232-233° Beilstein Reference: 2206277 UN No.: 3265 MDL No.: MFCD00000902
Formel: C₉H₁₁Cl
MW: 154,64 g/mol
Kokpunkt: 232…233 °C
Densitet: 1,050
Flampunkt: 65 °C (149 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000902
CAS nummer: 824-45-3
EINECS: 212-529-6
UN: 3265
ADR: 8,II