2,4,6-Trimethylbenzoic acid ≥99%
Leverantör: Thermo Fisher Scientific
A11831.09
A11831.18
A11831.30
A11831.09EA
1400
SEK
A11831.09
A11831.18
A11831.30
2,4,6-Trimethylbenzoic acid ≥99%
2,4,6-Trimethylbenzoic acid
Purity: 99%
CAS No.: 480-63-7
Molecular Formula: C10H12O2
Formula Weight: 164.20
Melting Point: 153-155°
Beilstein Reference: 1866187
Formel:
C₁₀H₁₂O₂ Kokpunkt: 298 °C (1013 hPa) Smältpunkt: 153…155 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00002481 CAS nummer: 480-63-7 EINECS: 207-553-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...