1,4-Diethylbenzene 98%
Leverantör: Thermo Scientific
Fara
114150050 114150250
ACRO114150050EA 1150 SEK
ACRO114150050 ACRO114150250
1,4-Diethylbenzene 98%
1,4-Diethylbenzene
Formel: C₁₀H₁₄
MW: 134,22 g/mol
Kokpunkt: 183…184 °C (1013 hPa)
Smältpunkt: –43 °C
Densitet: 0,862 g/cm³
Flampunkt: 56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009264
CAS nummer: 105-05-5
EINECS: 203-265-2
UN: 2049
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4935 to 1.4955 (20°C, 589 nm)
GC ≥97.5 %
UV (10 % in water)
Titration with NaOH ≥98.5 %
Membrane filtration test (10 % in water) Passes test
Suitability (HPLC gradient) Passes test